Grant Bio™ Densitometer
Grant Bio™ Densitometer
Actual product may vary
Fisherbrand
Grant Bio™ Densitometer
Measures turbidity of cell suspensions in a variety of life science applications. Grant Instruments™ Densitometer is designed and factory calibrated to measure turbidity in the range of 0.5 to 4.0 McFarland units (DEN-1) 0.0 to >7.5 McFarland units (DEN-1B) with a small standard deviation.
Have Questions?
Catalog NumberRangeDisplay Type
FKS114410780.3 to 15.0 McFarlandLED
Catalog number FKS11441078
Price (SGD)
3,471.00
EA
Add to cart
Range:
0.3 to 15.0 McFarland
Display Type:
LED
Price (SGD)
3,471.00
EA
Add to cart
  • Measurement range: 0.30 to 15.0 McFarland units (DEN-1), 0.0 to 6.0 McFarland units (DEN-1B)
  • Measurement time : 1 second
  • Precision: ±3%
  • Readings, conveniently shown in McFarland units, are clearly visible on the bright LED display
  • Factory calibrated - retains calibration without power supply
  • Can be user-calibrated with commercial standards or cell suspensions prepared in the laboratory
  • Designed for tubes with an outer diameter of 18mm
  • 16mm tubes can also be accommodated by using the optional tube adaptor D-16
  • Extremely compact design with small footprint and low profile that fits easily into the smallest workspace
  • DEN-1b - either battery powered by 3 x AAA or via external power supply.
Specifications
Light SourceLED
Display TypeLED
Sample Size≥2
Precision±3%
Range0.3 to 15.0 McFarland
Turbidity0.3 to 15.0 (McFarland units)
Dimensions (L x W x H)115 x 165 x 75 mm
Width (Metric)165 mm
Height (Metric)75 mm
Electrical RequirementsInput AC 120/230V 50/60 Hz Output DC 12 V
Voltage12 V
Temperature (Metric) Operating4°C to 40°C
Weight (Metric)0.7 kg
Warranty2 Years
Unit SizeEA