pH Temperature Electrode
pH Temperature Electrode
Actual product may vary
Fisherbrand
pH Temperature Electrode
Jenway™ pH Temperature Electrodes are used in conjunction with Jenway™ pH meters – 3510, 3520, 3540, 3340, 3345, and 4330.
Have Questions?
Catalog NumberMaterialDescription
FKS11440479GlassBulb end type
Catalog number FKS11440479
Price (SGD)
434.00
EA
Add to cart
Material:
Glass
Description:
Bulb end type
Price (SGD)
434.00
EA
Add to cart
  • Used in conjunction with Jenway™ pH meters – 3510, 3520, 3540, 3340, 3345, and 4330.
  • Combination Type
  • Refillable
  • Reference Type: AgCl
  • Isopotential: 0±20mV
Specifications
Connector TypesBNC
DescriptionBulb end type
Diameter (Metric)12 mm
Dimensions (L x W)12 dia. x 120 mm L
TypeCombination
For Use With (Application)General purpose pH measurement for accurate Q.C. and research work
Length (Metric)120 mm
MaterialGlass
pH Range0 to 14
Temperature (Metric) Range0°C to 80°C
Unit SizeEA