Stuart™ Hot Plate, Ambient to 375°C, Glass Ceramic
Stuart™  Hot Plate, Ambient to 375°C, Glass Ceramic
Actual product may vary
Fisherbrand
Stuart™ Hot Plate, Ambient to 375°C, Glass Ceramic
Shape: Rectangle; Size: 11.8 x 19.7 in.
Have Questions?
Catalog NumberTop Plate Material
FKS11436518Glass Ceramic
Catalog number FKS11436518
Price (SGD)
4,195.00
PK
Add to cart
Top Plate Material:
Glass Ceramic
Price (SGD)
4,195.00
PK
Add to cart
Features a glass ceramic top with excellent chemical resistance. The surface is easy to clean with high plate temperature settings for faster heating. Possesses an integral fitting for a retort rod and the bottom is shaped to allow a retort base to slide underneath the unit if required so saving further bench space.
  • Chemically resistant ceramic top plate
  • Flashing 'Hot' warning light, mains independent
  • Accurate temperature control with LED setting scale
  • Compact space saving design
  • IP rating 32
Specifications
Top Plate MaterialGlass Ceramic
Controller TypeKnob
Size11.8 x 19.7 in.
Temperature (Metric)Ambient to 375°C
Depth (Metric)360 mm
Height (Metric)130 mm
Weight (Metric)12 kg
Width (Metric)520 mm
Wattage2250 w
Electrical Requirements230 V, 50/60 Hz, 1200 w
Frequency50/60 Hz
ModelCB500
ShapeRectangle
Voltage230 V
TypeHot Plate
Unit SizePK