Stuart™ Digital Tube Roller
Stuart™ Digital Tube Roller
Actual product may vary
Fisherbrand
Stuart™ Digital Tube Roller
Provides a gentle, but highly efficient, rocking and rolling action. Stuart™ Digital Tube Rollers offer variable speed from 5 to 60rpm and a built-in digital timer which can either set from 1 second to 90 minutes, 1 minute to 9 hours or in 'minutes-only' mode up to 9999 minutes for maximum versatility.
Have Questions?
Catalog NumberIncludesDepth (Metric)
FKS114264986 Rollers255 mm
Catalog number FKS11426498
Price (SGD)
5,369.00
EA
Add to cart
Includes:
6 Rollers
Depth (Metric):
255 mm
Price (SGD)
5,369.00
EA
Add to cart
These roller mixers provide a gentle, but highly efficient, rocking and rolling action. The six roller model has a small space saving footprint, while the nine roller unit has a higher capacity. These advanced digital models offer variable speed from 5 to 60rpm and a built-in digital timer which can either set from 1 second to 90 minutes, 1 minute to 9 hours or in 'minutes-only' mode up to 9999 minutes for maximum versatility. Recommended for mixing blood samples, viscous substances and liquid-solid suspensions where minimum aeration is required. Units can be used in incubators up to 60°C and humidity up to 80%, or in cold rooms down to 4°C. Both roller mixers are robustly constructed and designed for easy cleaning, having plastic rollers and a drip tray to collect accidental spillages. Most sizes of tubes, Bijoux, Universals and bottles can be accommodated, up to 100mm diameter. An accessory stacking system SRT/STACK is available where bench space is at a premium.
  • Rocking and rolling action for complete mixing
  • Digital variable speed models with integrated timer
  • Six roller design with small footprint or nine roller model for high throughput
  • Designed for continuous or timed quiet operation
  • Can be used in cold rooms or in incubators, 4 to 60°C, 80% offrH
  • Gentle, highly efficient, rocking and rolling action.
  • Small space saving footprint
  • Variable speed from 5 to 60rpm
  • Built-in digital timer from 1 second to 90 minutes, 1 minute to 9 hours or in “minutes only” mode up to 9999 minutes for maximum versatility
  • Recommended for mixing blood samples, viscous substances and liquid-solid suspensions where minimum aeration is required
  • Most sizes of tubes, Bijoux, Universals and bottles can be accommodated, up to 100mm diameter
  • Accessory stacking system SRT/STACK is available where bench space is at a premium.
Specifications
Speed Range5 to 60 rpm
Electrical Requirements230 V, 50 Hz, 50 W
Voltage230 V
Wattage50 W
Frequency50 Hz
Depth (Metric)255 mm
Dimensions (L x W x H)340 x 565 x 110 mm
Height (Metric)110 mm
Length (Metric)340 mm
Width (Metric)565 mm
Controller TypeDigital
ModelSRT6D
For Use With (Application)Mixing blood samples, viscous substances and liquid-solid suspensions where minimum aeration is required
Humidity RangeUp to 80%
Includes6 Rollers
Packaging Quantity1 Ea.
Timer Range1 sec. to 9 hr.
Unit SizeEA