Thioacetamide Solution, EP Grade, Reagecon™
Thioacetamide Solution, EP Grade, Reagecon™
Thioacetamide Solution, EP Grade, Reagecon™
Thioacetamide Solution, EP Grade, Reagecon™
Fisherbrand
Thioacetamide Solution, EP Grade, Reagecon™
Manufactured according to Eur. Pharm. Chapter 4 (4.1.1) and prepared in an ISO 9001 facility. Thioacetamide Solution, EP Grade, Reagecon™ is a ready-to-use reagent with 40g/L of C2H5NS in water used to supply sulfide ions in manual and automated titrations.
Have Questions?
Catalog NumberQuantityPackaging
FKS114247491 LTamper evident
Catalog number FKS11424749
Price (SGD)
-
Quantity:
1 L
Packaging:
Tamper evident
Request bulk or custom format
Specifications
Chemical Name or MaterialThioacetamide
Shelf Life12 months
CAS62-55-5
CAS7732-18-5
ColorColorless
View more
Thioacetamide is an organosulfur compound that serves as a source of sulfide ions in the synthesis of organic and inorganic compounds. When a solution of thioacetamide is used to treat an aqueous solution of many metal cations, the corresponding metal sulfide results. This solution is prepared by adding 40g per 1L water.
  • Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4 (4.1.1) guidelines
  • Provided with a certificate of analysis and a label that shows the lot and expiration date
  • Ready to use product eliminates the time and labor of preparing precise reagents and volumetric solutions