Check Order Status
pH 12.00 Buffer Solution at 25°C, Reagecon
Element Search
Structure Search
Fisherbrand
pH 12.00 Buffer Solution at 25°C, Reagecon
Tested, certified, NIST traceable, and shelf-stable buffer solution.
Have Questions?
Catalog Number
Quantity
FKS11424569
500 mL
Catalog number
FKS11424569
Price (SGD)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Description
Accuracy ± 0.05 pH units
pH
12.00 ±0.05
Physical Form
Liquid
Quantity
500 mL
Packaging
HDPE Bottle GL45
View more
pH Buffer Reference Standard, pH Buffer Reference Material
Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
Guaranteed stability throughout its entire shelf life, even after opening the bottle.