Glass Bodied Conductivity Probe
Glass Bodied Conductivity Probe
Actual product may vary
Fisherbrand
Glass Bodied Conductivity Probe
Used in conjunction with Jenway™ Conductivity meters - 4510, 4520 and 3540. Jenway™ Glass Bodied Conductivity Probes are designed for general purpose conductivity, pure water, and low conductivity solutions.
Have Questions?
Catalog NumberCell ConstantMeasurement Ranges
FKS114105091 ±10%0 to 150 mS/cm
Catalog number FKS11410509
Price (SGD)
1,108.00
EA
Add to cart
Cell Constant:
1 ±10%
Measurement Ranges:
0 to 150 mS/cm
Price (SGD)
1,108.00
EA
Add to cart
  • For use Jenway™ conductivity meters – 4510, 4520 and 3540
  • Glass Bodied
  • Platinum plated
Specifications
For Use With (Application)General purpose conductivity measurement for accurate Q.C. and research work
For Use With (Equipment)4510, 4210, and 3540 Conductimeters
Temperature (Metric) Range0°C to 100°C
Cell Constant1 ±10%
TypeConductivity Cell
Diameter (Metric) Body12 mm
Length (Metric)120 mm
Length (Metric) Cable1 m
MaterialGlass Body, Platinum Plate
Measurement Ranges0 to 150 mS/cm
ProbeConductivity
Unit SizeEA