1x1 image pixel for data collection
MBL™ One Mark Class AS Glass Pipets
MBL™ One Mark Class AS Glass Pipets
Actual product may vary
Fisherbrand
MBL™ One Mark Class AS Glass Pipets
Manufactured from soda lime glass and complies with ISO 648 and DIN 12691. MBL™ One Mark Class A S Glass Pipets are calibrated for delivery and include markings in permanent amber stain.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ToleranceColorAccuracy
FKS1139964310 mL±0.02 mLRed±0.02 mL
FKS11349653100 mL±0.08 mLYellow±0.08 mL
FKS1132965325 mL±0.03 mLBlue±0.03 mL
FKS113896435 mL±0.015 mLWhite±0.015 mL
FKS1133965350 mL±0.05 mLRed±0.05 mL
Catalog number FKS11399643
Price (SGD)
43.00
PK
Add to cart
Volume (Metric):
10 mL
Tolerance:
±0.02 mL
Color:
Red
Accuracy:
±0.02 mL
Price (SGD)
43.00
PK
Add to cart
Manufactured from soda lime glass and complies with ISO 648 and DIN 12691. MBL™ One Mark Class A S Glass Pipets are calibrated for delivery and include markings in permanent amber stain.
  • Complies with ISO 648 and DIN 12691
  • Calibrated for delivery
  • Printed with batch code for downloadable batch certificate
  • Markings in permanent amber stain
  • Class AS with +5s waiting time
  • Manufactured from soda lime glass with robust tooled jets
Specifications
Accuracy±0.02 mL
CalibrationTo Deliver
Certifications/ComplianceISO 648, DIN 12691
ClassAS
MaterialSoda Lime Glass
GraduatedYes
Graduation ColorAmber
Quantity2 Pack
Tolerance±0.02 mL
ColorRed
Volume (Metric)10 mL
TypeBulb Pipet
Unit SizePK
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link