Sterilin™ 7mL Polystyrene Bijou Containers
Sterilin™ 7mL Polystyrene Bijou Containers
Actual product may vary
Fisherbrand
Sterilin™ 7mL Polystyrene Bijou Containers
Store small-volume samples in Thermo Scientific™ Sterilin™ 7mL Polystyrene Bijou Containers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityLabelIncludes
FKS11399133Non-sterilePrintedContainer Only
FKS11369123Non-sterileNo LabelContainer Only
Catalog number FKS11399133
Price (SGD)
896.00
PK
Add to cart
Sterility:
Non-sterile
Label:
Printed
Includes:
Container Only
Price (SGD)
896.00
PK
Add to cart
Store small-volume samples in Thermo Scientific™ Sterilin™ 7mL Polystyrene Bijou Containers.
NOT AVAILABLE IN AMERICA
Features:
  • Leak-tested in accordance with BS EN ISO 6717
  • Available pre-filled with boric acid (0.09g)
Specifications
Product TypeBijou Container
MaterialPolystyrene
ShapeCylindrical
SterilityNon-sterile
Capacity (Metric)7 mL
Certifications/ComplianceLeak-tested in accordance with BS EN ISO 6717
Closure MaterialPolyethylene
DescriptionBijou
Diameter (Metric)22.5 mm
Height (Metric)50.2 mm
IncludesContainer Only
LabelPrinted
No. per Case700/Cs.
Unit SizePK