Polyethylene Bags
Polyethylene Bags
Actual product may vary
Polyethylene Bags
Polyethylene Bags
Polyethylene Bags
Fisherbrand
Polyethylene Bags
Transparent and excellent for shipping, storage and as sample bags.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWidth (Metric)Height (Metric)Color
FKS1139820340 mm65 mmClear
FKS10031474125 mm190 mmClear
Catalog number FKS11398203
Price (SGD)
4.00
PK
Add to cart
Width (Metric):
40 mm
Height (Metric):
65 mm
Color:
Clear
Price (SGD)
4.00
PK
Add to cart
  • Available in a wide range of sizes/capacities
  • With resealable strips
Specifications
DescriptionResealable Bag
Capacity (Metric)30 mL
ColorClear
For Use With (Application)Ideal for storing stationery and sending samples
MaterialPolyethylene
Width (Metric)40 mm
Height (Metric)65 mm
Unit SizePK