Azlon™ Polypropylene Test Tube Rack
Azlon™ Polypropylene Test Tube Rack
Actual product may vary
Fisherbrand
Azlon™ Polypropylene Test Tube Rack
Economical and sturdy test tube racks supplied in self assembly flat packs
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorPositionsDiameter (Metric) OpeningDimensions (L x W x H)
FKS11397794White3226 mm265 x 126 x 88 mm
FKS11933849Blue2131 mm265 x 126 x 88 mm
Catalog number FKS11397794
Price (SGD)
261.00
PK
Add to cart
Color:
White
Positions:
32
Diameter (Metric) Opening:
26 mm
Dimensions (L x W x H):
265 x 126 x 88 mm
Price (SGD)
261.00
PK
Add to cart
Economical and sturdy test tube racks supplied in self assembly flat packs
  • Autoclavable at 121°C
  • Sinks in water baths
  • Alpha numeric place coding
Specifications
Product TypeTest Tube Rack
AutoclavableAutoclavable
ColorWhite
HoldsTubes (25 to 26 mm)
MaterialPolypropylene
Positions32
Dimensions (L x W x H)265 x 126 x 88 mm
Diameter (Metric) Opening26 mm
Unit SizePK