InLab™ Expert pH Electrodes
InLab™ Expert pH Electrodes
Actual product may vary
InLab™ Expert pH Electrodes
InLab™ Expert pH Electrodes
InLab™ Expert pH Electrodes
InLab™ Expert pH Electrodes
InLab™ Expert pH Electrodes
InLab™ Expert pH Electrodes
Fisherbrand
InLab™ Expert pH Electrodes
A combination pH electrode with PEEK plastic shaft for chemical and mechanical durability, and a fixed cable. Designed for use in a variety of applications.
Have Questions?
Catalog NumberConnector TypesIncludesLength (Metric) Cable
FKS11394203BNC, RCAISM Temperature Probe1.2 m
Catalog number FKS11394203
Price (SGD)
1,735.00
EA
Add to cart
Connector Types:
BNC, RCA
Includes:
ISM Temperature Probe
Length (Metric) Cable:
1.2 m
Price (SGD)
1,735.00
EA
Add to cart
Ideal for critical samples such as suspensions or samples of unknown composition. Mettler Toledo Expert Series pH and pH/ATC Combination Electrodes with open junction puts the reference electrode in direct contact with the measured medium, eliminating the possibility of contamination or blockage.
  • For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
  • Features a PEEK plastic shaft for harsh environments and requires no maintenance
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
  • General Lab
  • Beverages
  • Waste water
  • Suspended solids
  • Environmental
  • TRIS compatible
Accessories & Care
  • Open Junction and non-refillable - no additional electrolyte required
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
Connector TypesBNC, RCA
JunctionOpen
MaterialPEEK
pH Range0 to 14
Temperature (Metric) Operating0°C to 100°C
TypeCombination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology
Filling Solution TypeXerolyt Polymer
For Use With (Application)Emulsions, suspensions, effluent water and solutions containing Proteins and Sulphides
For Use With (Equipment)Fixed Cable to sensor
IncludesISM Temperature Probe
Internal ReferenceARGENTHAL
MembraneU Glass
ModelInLab Expert Pro-ISM
ProbeNTC 30 kilohm
Response Time<20 sec.
Temperature (English) Operating32°F to 212°F
Temperature (Metric) Range0°C to 100°C
Length (English)4.7 in.
Length (English) Cable47.2 in.
Length (Metric)120 mm
Length (Metric) Cable1.2 m
Diameter (English)0.47 in.
Diameter (Metric)12 mm
Unit SizeEA