Acrylic Pipette Stand
Acrylic Pipette Stand
Actual product may vary
Fisherbrand
Acrylic Pipette Stand
Leans back on an angle so there is no danger of pipettes falling out. Acrylic Pipette Stand grasps the pipettes in two places for secure hold.
Have Questions?
Catalog NumberColor
FKS11394045Clear
Catalog number FKS11394045
Price (SGD)
256.00
EA
Add to cart
Color:
Clear
Price (SGD)
256.00
EA
Add to cart
  • Acrylic construction.
  • Holds up to 6 pipettes.
  • Designed to work with most major pipette brands.
Specifications
Product TypePipette Stand
AutoclavableNot Autoclavable
ColorClear
HoldsPipetters
MaterialAcrylic
Positions6
ShapeRectangular
Shelves2 Tiers
StackableNo
SterilityNon-sterile
Quantity1 Ea.
Length (Metric)29.5 cm
Length (English)11.6 in.
Width (Metric)17.2 cm
Width (English)6.8 in.
Height (Metric)20.3 cm
Height (English)8 in.
Diameter (Metric) OpeningUpper: 25 mm; Lower: 20 mm
Unit SizeEA