Pyrex™ Amber Borosilicate Glass Class A Certified Volumetric Flask
Pyrex™ Amber Borosilicate Glass Class A Certified Volumetric Flask
Actual product may vary
Fisherbrand
Pyrex™ Amber Borosilicate Glass Class A Certified Volumetric Flask
Manufactured from borosilicate glass with amber coating on exterior surfaces to facilitate use with light sensitive solutions
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeToleranceHeight (Metric)
FKS1139365450 mL12/2±0.05 mL140 mm
FKS11303664100 mL14/23±0.08 mL170 mm
FKS1137365410 mL10/19±0.02 mL90 mm
FKS1138365425 mL10/19±0.03 mL110 mm
Catalog number FKS11393654
Price (SGD)
703.00
PK
Add to cart
Capacity (Metric):
50 mL
Closure Size:
12/2
Tolerance:
±0.05 mL
Height (Metric):
140 mm
Price (SGD)
703.00
PK
Add to cart
  • Calibrated to tolerances required by United States Pharmacopoeia (USP) which are tighter than those of ISO/DIN standards
  • Engraved with individual serial number
  • Supplied with certificate of calibration traceable to National standards
  • Conformity ‘H’ certified
  • Polyethylene stopper
  • White enamel inscriptions
Specifications
Certifications/ComplianceISO 1042, DIN 12664, USP, ISO/DIN, Conformity 'H' certified
Closure MaterialPolyethylene
ColorAmber
DescriptionWhite enamel inscriptions
Capacity (Metric)50 mL
MaterialBorosilicate Glass
Height (Metric)140 mm
Tolerance±0.05 mL
GraduatedYes
ClassA
Closure Size12/2
Unit SizePK