Check Order Status
Pyrex™ Glass Cylinder with White Gradations
Actual product may vary
Fisherbrand
Pyrex™ Glass Cylinder with White Gradations
Manufactured to Class B tolerances. The Pyrex Glass Cylinders with White Graduations has a robust hexagonal foot.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Graduations
FKS11393554
25 mL
0.5 mL
FKS11313564
100 mL
1 mL
FKS11383554
10 mL
0.2 mL
FKS11303564
50 mL
1 mL
4 Options
Catalog number
FKS11393554
Price (SGD)
92.00
PK
Add to cart
Capacity (Metric):
25 mL
Graduations:
0.5 mL
Price (SGD)
92.00
PK
Add to cart
Complies with ISO 4788
Manufactured to Class B tolerances
Robust hexagonal foot
White enamel graduations which remain visible when using dark colored solutions
Manufactured from chemically resistant borosilicate glass
Specifications
Base Type
Hexagonal
Scale Color
White
Shape
Round
Type
Graduated Cylinder
Tolerance
±0.5 mL
Material
Borosilicate Glass
Certifications/Compliance
BS 604, DIN 12680, ISO 4788
Class
B
Capacity (Metric)
25 mL
Height (Metric)
170 mm
Scale
White enamel
Graduations
0.5 mL