1x1 image pixel for data collection
Whatman™ Qualitative Filter Paper: Grade 1 Circles
Whatman™ Qualitative Filter Paper: Grade 1 Circles
Actual product may vary
Fisherbrand
Whatman™ Qualitative Filter Paper: Grade 1 Circles
Covers a wide range of laboratory applications and is frequently used for clarifying liquids. Whatman™ Qualitative Filter Paper: Grade 1 Circles are widely used standard grade filter papers for routine applications with medium retention and flow rates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Quantity
FKS1139280590 mm100/Pk.
FIS09-805-1B15 mm500/Pk.
FKS1019397130 mm400/Pk.
FIS09-805A42.5 mm100/Pk.
FKS1043389142.5 mm100/Pk.
FKS1135280547 mm100/Pk.
FKS1137280570 mm100/Pk.
FIS09-805-34585 mm100/Pk.
FKS1073861185 mm100/Pk.
FIS09-805E110 mm100/Pk.
FKS11302815110 mm100/Pk.
FKS10424081125 mm100/Pk.
FKS11322815150 mm100/Pk.
FKS10434271185 mm100/Pk.
FKS11342815240 mm100/Pk.
FIS09-805K270 mm100/Pk.
FKS10786262270 mm100/Pk.
FIS09-805L320 mm100/Pk.
FKS10134071320 mm100/Pk.
FKS1137039430 mm100/Pk.
FIS09-805-1D400 mm100/Pk.
Catalog number FKS11392805
Price (SGD)
52.00
PK
Estimated availability date 01-Oct-2025
Add to cart
Diameter (Metric):
90 mm
Quantity:
100/Pk.
Price (SGD)
52.00
PK
Add to cart
Covers a wide range of laboratory applications and is frequently used for clarifying liquids. Whatman™ Qualitative Filter Paper: Grade 1 Circles are widely used standard grade filter papers for routine applications with medium retention and flow rates.
These cellulose filters are used in qualitative analytical techniques to determine and identify materials and for clarifying liquids. 
  • Particle Retention: >11μm
  • Porosity: medium
  • Filtration speed: ASTM™, 40 sec.; Herzberg, 150 sec.
  • Surface: smooth
  • Separates precipitates such as lead sulfate, calcium oxalate (hot) and calcium carbonate
  • Used for soil analysis and seed testing procedures in the agricultural industry
  • Used to separate solid foodstuffs from associated liquid or extract liquid in the food industry
  • Atmospheric dust is collected from airflow and the stain-intensity measured 
  • Impregnated with a chromogenic reagent and color formation quantified by optical reflectance for gas detection
Specifications
Airflow Rate13sec./100mL/sq. in.
Basis Weight87 g/m2
DescriptionGrade 1 Qualitative Filter Paper Standard Grade, circle, 90 mm
FormatDisc
MaterialCellulose
Particle Retention>11 μm
Filtration Speed150 sec.
Flow Rate57mL/min. (water)
For Use With (Application)A widely used standard grade filter paper for routine applications with medium retention and flow rate. It covers a wide range of laboratory applications and is frequently used for clarifying liquids.
GradeGrade 1
PorosityMedium
Quantity100/Pk.
Thickness (Metric)180 μm
TypeQualitative Filter Paper
Diameter (English)3.54 in.
Diameter (Metric)90 mm
Unit SizePK
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link