High-Impact Trays
High-Impact Trays
Actual product may vary
Fisherbrand
High-Impact Trays
Manufactured from high impact polystyrene for strength and durability. Kartell™ High Impact Trays are useful for holding bottles, vials, and other individual items together.
Have Questions?
Catalog NumberCapacity (Metric)
FKS113912051.9 L
Catalog number FKS11391205
Price (SGD)
64.00
EA
Add to cart
Capacity (Metric):
1.9 L
Price (SGD)
64.00
EA
Add to cart
  • General containers for lab use
    • Shallow units: instrument trays or coasters
    • Deep units: slop trays or sand baths
    • Larger units can be used as pipette trays
  • Not autoclavable
  • Ridged feet for stability
  • Sloped sides
Specifications
Capacity (Metric)1.9 L
Dimensions (L x W x H)Interior: 380 x 270 x 10 mm; Exterior: 408 x 300 x 42 mm
Height (Metric)10 mm (Interior), 21 mm (Exterior)
Length (Metric)380 mm (Interior), 408 mm (Exterior)
MaterialPolystyrene
TypeTray
Width (Metric)270 mm (Interior), 300 mm (Exterior)
No. per Pack1 Piece
ShapeRectangular
Unit SizeEA