Provides an extremely clear and accurate visual pH value using four different segments of dye-impregnated indicator papers. Whatman™ pH Indicator Papers, CF Strips use dyes that are chemically bonded to the paper and cannot be leached into solution.
Have Questions?
Change view
Catalog Number
pH Range
FKS11390684
4.5 to 10
FIS09-876-17
0 to 14
FKS10377481
0 to 14
3 Options
Catalog number FKS11390684
Price (SGD)
103.00
PK
Add to cart
pH Range:
4.5 to 10
Price (SGD)
103.00
PK
Add to cart
Provides an extremely clear and accurate visual pH value using four different segments of dye-impregnated indicator papers. Whatman™ pH Indicator Papers, CF Strips use dyes that are chemically bonded to the paper and cannot be leached into solution.
Plastic supports that retain stiffness in liquids
Paper segments on each strip have chemically bonded dyes; can't leach into solution
Complementary indicator charts provide clear comparison
Full-range strips (0 to 14) graduated in 1pH units; wide-range strips (4.5 to 10pH) in 0.5pH units
Packs of 100; each strip is approximately 0.25 × 3 in. (6 × 80mm)
Specifications
pH Range4.5 to 10
FormatStrip
Graduations0.5 pH
IncludesPlastic supports, paper segments on each strip, complementary indicator charts