Pyrex™175mm Diameter Glass Büchner Filter Funnel
Pyrex™175mm Diameter Glass Büchner Filter Funnel
Actual product may vary
Fisherbrand
Pyrex™175mm Diameter Glass Büchner Filter Funnel
Manufactured from chemically resistant Pyrex™ glass. Pyrex™ 175mm Diameter Glass Büchner Filter Funnel with integral sintered glass disc is highly resistant to most corrosive reagents and eliminates the need for filter papers.
Have Questions?
Catalog NumberPorosityPore Size
FKS11388004316 to 40μm
Catalog number FKS11388004
Price (SGD)
1,359.00
Each
Add to cart
Porosity:
3
Pore Size:
16 to 40μm
Price (SGD)
1,359.00
Each
Add to cart
  • Comprehensive range of Büchner filter funnels with integral sintered glass discs
  • Disc and funnel body manufactured from chemically resistant Pyrex™ glass
  • For convenient filtration of solutions
    - No filter papers required
    - Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Choice of disc diameters and porosities
Specifications
Capacity (Metric)4000 mL
Diameter (Metric) Stem30mm
MaterialBorosilicate Glass, Borosilicate Glass
Height (Metric)390 mm
For Use With (Application)For easy filtering solutions
Diameter (Metric) Disc175mm
IncludesIntegrated filter plate sintered Glass disc
Length (Metric) Stem150mm
Porosity3
Pore Size16 to 40μm
Product TypeBuchner Funnel
Unit SizeEach