Class A Reusable PMP Volumetric Flasks with Tapered PP Stopper
Class A Reusable PMP Volumetric Flasks with Tapered PP Stopper
Actual product may vary
Fisher Catalogue
Class A Reusable PMP Volumetric Flasks with Tapered PP Stopper
Autoclaved to 121°C
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS1138787150 mL
FIS10-210-63950 mL
FIS10-210-63710 mL
FIS10-210-640100 mL
FKS113578711 L
FIS10-210-642500 mL
Catalog number FKS11387871
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
50 mL
Translucent flask is supplied with tapered polypropylene stopper with an individually calibrated ­graduation line. The product is manufactured to Class A tolerances as defined by ISO 1042. They are supplied with an imprinted lot number and certificate. These flasks may be autoclaved to 121°C without affecting the graduation tolerance. Recommended routine cleaning up to 60°C will preserve graduation markings. Height dimension is for flask only, without stopper.
Specifications
Capacity (English)1.69 oz.
Closure MaterialPolypropylene
ColorTranslucent
DescriptionIndividually calibrated graduation line
Diameter (English) Outer2 in.
Capacity (Metric)50 mL
MaterialPMP
Height (English)5.5 in.
Height (Metric)14 cm
IncludesImprinted lot number and certificate
No. per Case6/Cs.
No. per Pack1/Pk.
Standard Taper Joint12/21
Temperature Range (English)140°F
Temperature Range (Metric)60°C
Tolerance0.06 mL
AutoclavableAutoclavable
DisposableReusable
Diameter (Metric) Outer51 mm
Unit SizePK