Azlon™ Industrial HDPE Funnel
Azlon™ Industrial HDPE Funnel
Actual product may vary
Fisher Catalogue
Azlon™ Industrial HDPE Funnel
Constructed of Polyethylene and unbreakable in normal use. Azlon™ Industrial HDPE Funnels are suitable for food contact applications in accordance with EU and USA regulations. Feature straight sided upper rims to prevent spillage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)TopDiameter (Metric) StemHeight (Metric)
FKS11383664180mm17mm181 mm
FKS10504355290mm27mm277 mm
Catalog number FKS11383664
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric)Top:
180mm
Diameter (Metric) Stem:
17mm
Height (Metric):
181 mm
Constructed of Polyethylene and unbreakable in normal use. Azlon™ Industrial HDPE Funnels are suitable for food contact applications in accordance with EU and USA regulations. Feature straight sided upper rims to prevent spillage.
  • Rigid industrial funnels in polyethylene
  • Unbreakable in normal use
  • Feature straight sided upper rims to prevent spillage
Specifications
Diameter (Metric)Top180mm
Capacity (Metric)1500 mL
Diameter (Metric) Stem17mm
MaterialHDPE
Height (Metric)181 mm
Unit SizePK