Pyrex™ Borosilicate Glass Conical Beakers
Pyrex™  Borosilicate Glass Conical Beakers
Actual product may vary
Fisherbrand
Pyrex™ Borosilicate Glass Conical Beakers
Use with a wide variety of media and liquids. Pyrex™ borosilicate glass conical beakers are versitle for a wide range of laboratory applications
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS11383464250 mL
FKS11393464500 mL
Catalog number FKS11383464
Price (SGD)
451.00
Pack
Add to cart
Capacity (Metric):
250 mL
Price (SGD)
451.00
Pack
Add to cart
Use with a wide variety of media and liquids. Pyrex™borosilicate glass conical beakers are versitle for a wide range of laboratory applications
  • Conical shape (Phillips pattern) with spout
  • Manufactured from Pyrex™borosilicate glass
  • Excellent thermal performance and chemical resistance
Specifications
Capacity (Metric)250 mL
MaterialBorosilicate Glass
Height (Metric)105 mm
Quantity10/Pk.
TypeBeaker
Unit SizePack