Quickfit™ Borosilicate Glass Soxhlet Extractor
Quickfit™ Borosilicate Glass Soxhlet Extractor
Actual product may vary
Fisherbrand
Quickfit™ Borosilicate Glass Soxhlet Extractor
Conforms to BS 2071. Quickfit™ Borosilicate Glass Soxhlet Extractor removes soluble components or impurities from solids via repeated soaking/siphoning with solvent. A wide choice of extractors is available to accommodate most sizes of sample and thimbles. Accurately formed concentric siphon tube provides reliable performance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Cone SizeStandard Taper Joint Bottom
FKS11381885200 mL250 mm24/2950/42
FKS11371885100 mL220 mm24/2940/38
Catalog number FKS11381885
Price (SGD)
634.00
EA
Add to cart
Capacity (Metric):
200 mL
Height (Metric):
250 mm
Cone Size:
24/29
Standard Taper Joint Bottom:
50/42
Price (SGD)
634.00
EA
Add to cart
Conforms to BS 2071. Quickfit™ Borosilicate Glass Soxhlet Extractor removes soluble components or impurities from solids via repeated soaking/siphoning with solvent. A wide choice of extractors is available to accommodate most sizes of sample and thimbles. Accurately formed concentric siphon tube provides reliable performance.
  • Conforms to BS 2071
  • For extracting soluble components or impurities from solids via repeated soaking/siphoning with solvent
  • Wide choice of extractors to accommodate most sizes of sample and thimbles
  • Accurately formed concentric siphon tube for reliable performance
Specifications
Product TypeSoxhlet Extractor
Capacity (Metric)200 mL
MaterialPyrex Borosilicate Glass
Cone Size24/29
For Use With (Application)Ideal for extracting soluble components or impurities from solids via repeated soaking/siphoning with solvent
Certifications/ComplianceBS 2071
Standard Taper Joint Bottom50/42
Height (Metric)250 mm
Unit SizeEA