Translucent Polypropylene Volumetric Flasks with Screw Cap
Translucent Polypropylene Volumetric Flasks with Screw Cap
Actual product may vary
Fisherbrand
Translucent Polypropylene Volumetric Flasks with Screw Cap
Accurately measure precise volumes of a liquid. Kartell™ Translucent Polypropylene Volumetric Flasks with Screw Cap is used for dilutions and to prepare chemical solutions.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1138122550 mL
Catalog number FKS11381225
Price (SGD)
34.00
EA
Add to cart
Capacity (Metric):
50 mL
Price (SGD)
34.00
EA
Add to cart
Accurately measure precise volumes of a liquid. Kartell™ Translucent Polypropylene Volumetric Flasks with Screw Cap is used for dilutions and to prepare chemical solutions.
  • Create no meniscus so final readings are easy
  • Calibrated gravimetrically at 20°C
  • Translucency of PP enhanced when in contact with liquid
  • Thick walls eliminate distortion
  • Thin neck make final reading precise
  • Autoclavable
  • Chemically non absorbant
  • No wetting
  • Screw caps included
  • Suitable for foodstuff
Specifications
ColorTranslucent
Capacity (Metric)50 mL
MaterialPolypropylene
Height (Metric)143 mm
IncludesScrew Caps
Neck StyleGL18
Tolerance±1 mL
AutoclavableAutoclavable
Closure TypeScrew Cap
Diameter (Metric) Outer46 mm
Unit SizeEA