Whatman™ Binder-Free Glass Microfiber Filters, Grade 934-AH Circles
Whatman™ Binder-Free Glass Microfiber Filters, Grade 934-AH Circles
Actual product may vary
Fisherbrand
Whatman™ Binder-Free Glass Microfiber Filters, Grade 934-AH Circles
Designed for analysis of non-filterable residue and materials that require a high loading capacity glass microfiber filter. Whatman™ Binder-Free Glass Microfiber Filters, Grade 934-AH Circles are also used in cell harvesting, liquid scintillation, and monitoring pollution.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
FKS1138061455 mm
FIS09-873B24 mm
FKS1047160224 mm
FIS09-873DD47 mm
FKS1137061447 mm
FKS1002217190 mm
FIS09-873J125 mm
Catalog number FKS11380614
Price (SGD)
426.00
PK
Add to cart
Diameter (Metric):
55 mm
Price (SGD)
426.00
PK
Add to cart
Designed for analysis of non-filterable residue and materials that require a high loading capacity glass microfiber filter. Whatman™ Binder-Free Glass Microfiber Filters, Grade 934-AH Circles are also used in cell harvesting, liquid scintillation, and monitoring pollution.
Grade 934-AH filters are often used in total suspended solids analysis.
  • Smooth surface for efficient precipitate recovery
  • Nominal particle retention of 1.5μm
  • Withstands temperatures up to 550°C
  • Fast filtration speed
Specifications
Basis Weight64 g/m2
DescriptionWhatman Grade 934-AH borosilicate glass microfiber filters from GE are smooth water quality filters with high retention efficiency. Grade 934-AH filters are often used in total suspended solids analysis.
FormatDisc
MaterialBorosilicate Glass
Particle Retention1.5 μm
Filtration Time47 sec./100mL
Flow Rate3.7 sec./100mL/in.2
GradeGrade 934-AH
Quantity100 Pack
ShapeCircle
Surface TypeSmooth
Temperature (English) Max.932°F
Temperature (Metric) Max500°C
Thickness (English)0.017 in.
Thickness (Metric)435 μm
TypeFilter for Total Suspended Solids Analysis
Waterflow Rate341 mL/min
Diameter (English)2.1 in.
Diameter (Metric)55 mm
Unit SizePK