Manufactured from polypropylene for superior chemical and thermal stability. Whatman™ UNIPLATE™ Microplates are suitable for simple filtrate collection and homogeneous assay techniques used in high-throughput screening (HTS).
Have Questions?
Catalog Number
Sterility
Volume (Metric) Well
Well Shape
No. of Wells
FKS11378674
Non-sterile
10 mL
Round
24
Catalog number FKS11378674
Price (SGD)
2,025.00
PK
Add to cart
Sterility:
Non-sterile
Volume (Metric) Well:
10 mL
Well Shape:
Round
No. of Wells:
24
Price (SGD)
2,025.00
PK
Add to cart
Manufactured from polypropylene for superior chemical and thermal stability. Whatman™ UNIPLATE™ Microplates are suitable for simple filtrate collection and homogeneous assay techniques used in high-throughput screening (HTS).
Withstand low temperatures (-80°C) for long-term storage
Conform to ANSI™/SBS standards for compatibility with most microplate readers and automated plate-handling devices
Specifications
FormatStandard
DescriptionUNIPLATE microplates
Array4 x 6
ColorNatural
MaterialPolypropylene
Volume (Metric) Well10 mL
No. of Wells24
SterilityNon-sterile
Well ShapeRound
For Use With (Application)Ideal for homogeneous assay techniques utilized in high-throughput screening