Round Lead Rings With Vikem™ Vinyl Coating
Round Lead Rings With Vikem™ Vinyl Coating
Actual product may vary
Round Lead Rings With Vikem™ Vinyl Coating
Round Lead Rings With Vikem™ Vinyl Coating
Fisherbrand
Round Lead Rings With Vikem™ Vinyl Coating
Protects and stabilizes expensive glassware.Lead rings are safe for use with fragile laboratory glassware.
Have Questions?
Catalog number FKS11377174
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Protects and stabilizes expensive glassware.Lead rings are safe for use with fragile laboratory glassware.
  • Stabilize glassware immersed in water baths
  • Reduce floating, breakage and chemical spills
  • Chemical-resistant
  • Will not scratch flasks
Specifications
Product TypeLead Ring
AutoclavableNot Autoclavable
ColorBright Red
HoldsFlasks (250 to 1000 mL)
MaterialLead
ShapeRing
ShelvesNone
StackableYes, When Empty
SterilityNon-sterile
For Use With (Equipment)250 to 1000mL FlaskmL
Length (Metric)89 mm
Width (Metric)89 mm
Height (Metric)15.9 mm
Diameter (English) Inner2 in.
Diameter (English) Outer3.5 in.
Diameter (Metric) Outer8.9 cm
Diameter (Metric) Inner5.1 cm
Weight (English)1 lb.
Weight (Metric)454 g
Unit SizeEA