PCR Tubes with Flat Cap, 0.2 mL
PCR Tubes with Flat Cap, 0.2 mL
Actual product may vary
Fisherbrand
PCR Tubes with Flat Cap, 0.2 mL
Designed to fit standard 96-well blocks. Axygen™ PCR Tubes with Flat Cap, 0.2m are constructed of non-sterile polypropylene. Each conical tube features thin and consistent wall thickness. Tight-fitting cap secures contents and prevents evaporation. Available in an assortment of colors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorProduct Type
FKS11376044ClearThin Wall PCR Tube
FIS14-222-260AssortedThin Wall PCR Tube
FIS14-222-262ClearThin Wall PCR Tube
FIS14-222-283WhiteMaxymum Recovery PCR Tube
Catalog number FKS11376044
Price (SGD)
227.00
PK
Add to cart
Color:
Clear
Product Type:
Thin Wall PCR Tube
Price (SGD)
227.00
PK
Add to cart
Designed to fit standard 96-well blocks. Axygen™ PCR Tubes with Flat Cap, 0.2m are constructed of non-sterile polypropylene. Each conical tube features thin and consistent wall thickness. Tight-fitting cap secures contents and prevents evaporation. Available in an assortment of colors.
  • Quantitative PCR compatible tubes for a wide range of real-time PCR instruments
  • Ultra-thin and consistent wall thickness allows precise thermal transfer
  • Even insertion cap seals full length for a tight fit that prevents sample evaporation
  • Maxymum Recovery™ option provides ultra-low retention and high yield
  • Certified RNase-/DNase-free and nonpyrogenic
  • Manufactured with 99.9% pure virgin polypropylene
  • Available in a wide variety of colors to support color-coding
Specifications
Certifications/ComplianceRNase, DNase, pyrogen-free
ColorClear
Volume (Metric)0.2 mL
Quantity1000/Pk., 10Pk./Cs.
Closure MaterialPolypropylene
For Use With (Equipment)Standard 96-well blocks of real-time PCR instruments and thermal transfer applications
ShapeConical
SterilityNon-sterile
MaterialPolypropylene
Closure TypeFlat Cap
No. of Wells96
FormatTube
Product TypeThin Wall PCR Tube
Unit SizePK