Whatman™ HEPA-Vent Filter
Whatman™ HEPA-Vent Filter
Actual product may vary
Whatman™ HEPA-Vent Filter
Whatman™ HEPA-Vent Filter
Fisherbrand
Whatman™ HEPA-Vent Filter
Strengthened glass filter media by dual lamination with a tough polyester monofilament. Whatman™ HEPA-Vent Filters are ideal for air and gas filtration applications where high retention, dirt-holding capacity and flow rates are required.
Have Questions?
Catalog NumberDiameter (Metric)
FKS1137463450 mm
Catalog number FKS11374634
Price (SGD)
523.00
PK
Add to cart
Diameter (Metric):
50 mm
Price (SGD)
523.00
PK
Add to cart
Strengthened glass filter media by dual lamination with a tough polyester monofilament. Whatman™ HEPA-Vent Filters are ideal for air and gas filtration applications where high retention, dirt-holding capacity and flow rates are required.
  • Laminated hydrophobically treated glass microfiber
  • Preventing bacterial, algal or fungal contamination in fermentors or incubators
  • Retains 99.97% of all particles ≥0.3μm in air
  • Autoclavable
  • Bidirectional flow
  • Filter area: 16cm2
  • Depth filter design allows for high loading capacity
  • Tissue culture applications
  • Heat-fused sealing
Specifications
AutoclavableAutoclavable
DescriptionHEPA-VENT 50 mm Disc, inlet 1/4 to 3/8 in. stepped barb, outlet 1/4 to 3/8 in. stepped barb (10 pcs)
Material (Membrane)Glass Fiber
Filtration Area16 cm2
Housing MaterialPolypropylene
Inlet Connection (Metric)1/4 to 3/8 in SB
Inlet Style1/4 to 3/8 in. stepped barb
Max. Pressure4.1 bar - Capsule (60 psi)
Outlet Style1/4 to 3/8 in. stepped barb
Quantity10 Pieces
TypeVent Filter
Diameter (Metric)50 mm
Diameter (Metric) Disc50 mm
Unit SizePK