PYREX™ Pear-Shaped Glass Separating Funnel with PTFE Key
PYREX™ Pear-Shaped Glass Separating Funnel with PTFE Key
Actual product may vary
Fisherbrand
PYREX™ Pear-Shaped Glass Separating Funnel with PTFE Key
Manufactured from Pyrex™ glass for corrosion-free performance. Pyrex™ Pear-Shaped Glass Separating Funnel with Glass Key complies with ISO 4800 Type 2 Standards. Includes glass stopper and interchangeable PTFE key.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Stopcock
FKS11373684500 mL4mm; Glass
FKS11363684250 mL2.5mm; Glass
Catalog number FKS11373684
Price (SGD)
457.00
EA
Add to cart
Capacity (Metric):
500 mL
Stopcock:
4mm; Glass
Price (SGD)
457.00
EA
Add to cart
Manufactured from Pyrex™ glass for corrosion-free performance. Pyrex™ Pear-Shaped Glass Separating Funnel with Glass Key complies with ISO 4800 Type 2 Standards. Includes glass stopper and interchangeable PTFE key.
  • Complies with ISO 4800 Type 2
  • Manufactured from Pyrex™ glass for corrosion-free performance
  • With interchangeable PTFE key
  • Supplied with glass stopper
Specifications
Capacity (Metric)500 mL
MaterialBorosilicate Glass, Borosilicate Glass
Certifications/ComplianceISO 4800
Quantity1PK
Stopcock4mm; Glass
Stopper MaterialGlass
Stopper Number24/29
Product TypeSeparatory Funnel
Unit SizeEA