Pyrex™ Amber Borosilicate Glass Class A Volumetric Flask
Pyrex™ Amber Borosilicate Glass Class A Volumetric Flask
Actual product may vary
Fisher Catalogue
Pyrex™ Amber Borosilicate Glass Class A Volumetric Flask
Manufactured from Pyrex™ Borosilicate Glass with an amber coating on exterior surfaces to facilitate use with light sensitive solutions. Pyrex™ Borosilicate Glass Class A Volumetric Flask complies with ISO 1042 and DIN 12664 and confirmity 'H' certified. Calibrated to the tolerance required by the United States Pharmacopoeia (USP.) Each flask is laser etched with an individual serial number batch code and date of manufacture.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeHeight (Metric)Tolerance
FKS113736445 mL10/1970 mm±0.02 mL
FKS11313654100 mL14/23170 mm±0.08 mL
FKS1130365450 mL12/2140 mm±0.05 mL
Catalog number FKS11373644
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
5 mL
Closure Size:
10/19
Height (Metric):
70 mm
Tolerance:
±0.02 mL
Manufactured from Pyrex™ Borosilicate Glass with an amber coating on exterior surfaces to facilitate use with light sensitive solutions. Pyrex™ Borosilicate Glass Class A Volumetric Flask complies with ISO 1042 and DIN 12664 and is conformity 'H' certified. Calibrated to the tolerance required by the United States Pharmacopoeia (USP.) Each flask is laser etched with an individual serial number batch code and date of manufacture.
  • Complies with ISO 1042 and DIN 12664
  • Calibrated to the tolerance required by the United States Pharmacopoeia (USP) which are tighter than those of ISO/DIN standards
  • Manufactured from borosilicate glass with an amber coating on exterior surfaces to facilitate use with light sensitive solutions
  • Each flask is laser etched with an individual serial number batch code and date of manufacture
  • Conformity 'H' certified
  • Batch certificate available to download online
Specifications
Certifications/ComplianceISO 1042, DIN 12664, USP, Conformity 'H' certified
Closure MaterialPolyethylene
ColorAmber
DescriptionWhite enamel inscriptions
Capacity (Metric)5 mL
MaterialBorosilicate Glass
Height (Metric)70 mm
Tolerance±0.02 mL
GraduatedYes
ClassA
Closure Size10/19
Unit SizePK