Pyrex™ Borosilicate Glass Cylinder with Polyethylene Foot
Pyrex™ Borosilicate Glass Cylinder with Polyethylene Foot
Actual product may vary
Fisherbrand
Pyrex™ Borosilicate Glass Cylinder with Polyethylene Foot
Manufactured from Pyrex™ borosilicate glass and polyethylene for strength, stability and superior chemical resistance. Pyrex™ Borosilicate Glass Cylinder with Polyethylene Foot is an excellent device for measuring and mixing liquids in a laboratory.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)GraduationsHeight (Metric)
FKS11373564250 mL2 mL320 mm
FKS1135356450 mL1 mL195 mm
FKS11363564100 mL1 mL270 mm
Catalog number FKS11373564
Price (SGD)
171.00
PK
Add to cart
Capacity (Metric):
250 mL
Graduations:
2 mL
Height (Metric):
320 mm
Price (SGD)
171.00
PK
Add to cart
Manufactured from Pyrex™ borosilicate glass and polyethylene for strength, stability and superior chemical resistance. Pyrex™ Borosilicate Glass Cylinder with Polyethylene Foot is an excellent device for measuring and mixing liquids in a laboratory.
  • Borosilicate glass cylinder: white enamel graduations for optimal visibility
  • Polyethylene foot: absorbs impact and reduces breakage
  • Class B
  • Complies with: DIN 12680, BS 604, ISO 4788
  • Replacement feet: available separately
Specifications
Base TypeHexagonal
Scale ColorWhite
ShapeRound
TypeGraduated Cylinder
Tolerance±2 mL
MaterialBorosilicate Glass
Certifications/ComplianceBS 604, DIN 12680, ISO 4788
ClassB
Capacity (Metric)250 mL
Height (Metric)320 mm
IncludesPolyethylene Foot
ScaleWhite enamel
Graduations2 mL
Unit SizePK