QIAcard Bloodstain
QIAcard Bloodstain
Actual product may vary
Fisherbrand
QIAcard Bloodstain
For collection, stabilization, processing, transport and archiving of biological samples. 4 sample areas per card.
Have Questions?
Catalog NumberQuantity
FKS11373195100 Pk.
Catalog number FKS11373195
Price (SGD)
1,402.00
EA
Add to cart
Quantity:
100 Pk.
Price (SGD)
1,402.00
EA
Add to cart
For collection, stabilization, processing, transport and archiving of biological samples. 4 sample areas per card.
  • Non-Indicating and Non-FTA treated card designed for the collection, short-term handling of specimens.
  • Unlike FTA Cards, the Bloodstain Cards are not coated with a protective chemical cocktail and DNA is as risk of degradation and damage.
  • Convenient transport of blood and bodily fluids with protective cover included.
Specifications
FormatCard
For Use With (Application)Collection and transport of blood and body fluids
Quantity100 Pk.
Sample Volume125μL per sample area
Unit SizeEA