Check Order Status
Quickfit™ Allihn Condenser with Ground Cone
Actual product may vary
Fisherbrand
Quickfit™ Allihn Condenser with Ground Cone
Ideal for reflux applications.
Have Questions?
Catalog Number
Standard Taper Joint No
FKS11371845
50/42
Catalog number
FKS11371845
Price (SGD)
873.00
EA
Add to cart
Standard Taper Joint No:
50/42
Price (SGD)
873.00
EA
Add to cart
Ideal for reflux applications.
Inner bulb sections provide increased cooling surface
With detachable plastic screwthread connectors for easy and safe attachment of flexible tubing
Specifications
For Use With (Application)
Ideal for reflux work
Length (Metric)
365 mm
Length (Metric) Jacket
365 mm
Standard Taper Joint No
50/42
Material
Borosilicate Glass
Type
Allihn Condenser