Whatman™ SG81 Grade Chromatography Paper
Whatman™ SG81 Grade Chromatography Paper
Actual product may vary
Fisherbrand
Whatman™ SG81 Grade Chromatography Paper
SG81 ion exchange sheets is a 0.27 mm combining cellulose and large pore silica gel for separations in which both partition and adsorption are important.
Have Questions?
Catalog NumberLength (Metric)
FKS1137074457 cm
Catalog number FKS11370744
Price (SGD)
1,039.00
PK
Add to cart
Length (Metric):
57 cm
Price (SGD)
1,039.00
PK
Add to cart
  • Simultaneous development of multiple samples on the same sheet under identical conditions.
  • Sequential development of the same samples with different solvents and/or different concentrations of the same solvent.
  • Suitable for two-dimensional chromatography (change in direction of the solvent front) with possible improved resolution.
  • Used in enzyme assays to separate product from reactant by charge or polarity.
Specifications
Film Thickness0.27 mm
For Use With (Application)For Enzyme Assays to Separate Product From Reactant by Charge or Polarity
FormatSheet
GradeSG81
MaterialCellulose, Large Pore Silica
Width (English)18.1 in.
Width (Metric)46 cm
Flow Rate110 mm/30 min.
Length (Metric)57 cm
Unit SizePK