Whatman™ Quantitative Filter Papers: Grade 542 Circles
Whatman™ Quantitative Filter Papers: Grade 542 Circles
Actual product may vary
Fisherbrand
Whatman™ Quantitative Filter Papers: Grade 542 Circles
Enables high retention of fine particles in chemically harsh conditions. Whatman™ Quantitative Filter Papers: Grade 542 Circles are well suited for use in capturing fine precipitates in applications such as metal determinations and hydroxide collection.
Have Questions?
Catalog NumberDiameter (Metric)
FKS11370534125 mm
Catalog number FKS11370534
Price (SGD)
274.00
PK
Add to cart
Diameter (Metric):
125 mm
Price (SGD)
274.00
PK
Add to cart
Enables high retention of fine particles in chemically harsh conditions. Whatman™ Quantitative Filter Papers: Grade 542 Circles are well suited for use in capturing fine precipitates in applications such as metal determinations and hydroxide collection.
Whatman hardened ashless quantitative filter papers are acid-hardened to give high wet strength and chemical resistance with extremely low ash content. They are suitable for a wide range of critical filtration procedures.
  • High retention of fine precipitates in applications such as metal determinations and hydroxide collection
  • Collection of trace elements and radionuclides from the atmosphere
  • Nominal particle retention rating of 2.7 μm
  • Herzberg filtration speed of 2510 s
  • Extremely low ash content (0.006% ash maximum)
Specifications
Airflow Rate64 s/100 mL/in2
DescriptionGrade 542 Ashless Fine Precipitate FIlter, 125 mm circles (100 pcs)
Filter TypeHardened Ashless
FormatDisc
MaterialCotton Linters
Particle Retention2.7 μm
Filtration SpeedASTM, 250 seconds; Herzberg, 2510 seconds
For Use With (Application)Used in gravimetric metal determinations.
GradeGrade 542
PorosityFine
Quantity100/Pk.
Thickness (Metric)150 μm
TypeQuantitative Filter Paper
Diameter (English)4.92 in.
Diameter (Metric)125 mm
Unit SizePK