Nickel gauze, 100 mesh woven from 0.1mm (0.004in) dia wire
Nickel gauze, 100 mesh woven from 0.1mm (0.004in) dia wire
Nickel gauze, 100 mesh woven from 0.1mm (0.004in) dia wire
Nickel gauze, 100 mesh woven from 0.1mm (0.004in) dia wire
Fisherbrand
Nickel gauze, 100 mesh woven from 0.1mm (0.004in) dia wire
Form: Wire Cloth; Open area: 36%; Width of opening: 0.006 in. | CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Catalog NumberQuantity
FKS1136960875 x 75 mm
Catalog number FKS11369608
Price (SGD)
-
Quantity:
75 x 75 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialNickel
ColorSilver to Gray
CAS7440-02-0
Health Hazard 1H317-H351-H372
AppearanceSilver gray guaze