Sterilin™ Loops and Spreaders
Actual product may vary
Fisherbrand
Sterilin™ Loops and Spreaders
Simplify your culture work with Thermo Scientific™ Sterilin™ Loops and Spreaders. Choices for inoculation, dilution streaking, spreading and colony picking.
Have Questions?
Catalog NumberVolumeProduct TypeColorQuantity
FKS1136928310 μLInoculating Loop and Needle, RigidDark blue10
Catalog number FKS11369283
Price (SGD)
353.00
Pack
Volume:
10 μL
Product Type:
Inoculating Loop and Needle, Rigid
Color:
Dark blue
Quantity:
10
Price (SGD)
353.00
Pack
Simplify your culture work with Thermo Scientific™ Sterilin™ Loops and Spreaders. Choices for inoculation, dilution streaking, spreading and colony picking.
NOT AVAILABLE IN AMERICA
  • For inoculation, dilution streaking, spreading and picking isolated colonies
  • Eliminate the need for flaming and the subsequent risk of aerosols Inoculation loop for fixed sample volumes with choice of sphere for dilution streaking, or needle for picking colonies and stab cultures
  • Choice of rigid or flexible loops to suit different applications and user preferences
  • Hexagonal shaft improves grip and assists orientation
  • Free of lubricants, oils and electrostatic charges to facilitate consistent wetting and complete liquid transfer
  • No rough edges means smooth, problem-free plating and streaking of cultures
  • Sterilized by gamma irradiation
Ordering Information
  • Not available in all geographic regions
Specifications
SterilityIrradiated
Volume10 μL
ColorDark blue
Product TypeInoculating Loop and Needle, Rigid
Quantity10
Unit SizePack