Search Thermo Fisher Scientific
Catalog Number | Pore Size | Diameter (Metric) | Includes |
---|---|---|---|
FKS11364724 | 0.02 μm | 47 mm | Support Ring |
FIS09-926-36 | 0.1 μm | 47 mm | Support Ring |
FIS09-926-300 | 0.2 μm | 47 mm | No Support Ring |
FKS11345044 | 0.02 μm | 47 mm | No Support Ring |