1000 mL Vacuum Filter/Storage Bottle System
1000 mL Vacuum Filter/Storage Bottle System
Actual product may vary
Fisherbrand
1000 mL Vacuum Filter/Storage Bottle System
Vacuum filtration systems use a square filter membrane support system that increases filtration rates with printed funnels to identify membrane type and product number. The system consists of a polystyrene filter funnel joined by an adapter ring to a removable polystyrene storage bottle with a separate sterile polyethylene cap.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterial (Membrane)Funnel CapacityPore Size
FKS11363785PES1000 mL0.1 μm
CRG431474PES1000 mL0.1 μm
FIS09-761-180PES1000 mL0.1 μm
CRG430517Cellulose Acetate1000 mL0.22 μm
FIS09-761-40Cellulose Acetate1000 mL0.22 μm
CRG431098PES1000 mL0.22 μm
FIS09-761-108PES1000 mL0.22 μm
FKS10296191PES1000 mL0.22 μm
FIS09-761-104Cellulose Nitrate1000 mL0.22 μm
FIS09-761-41Cellulose Acetate1000 mL0.45 μm
FKS10730693Cellulose Acetate1000 mL0.45 μm
Catalog number FKS11363785
Price (SGD)
1,458.00
PK
Add to cart
Material (Membrane):
PES
Funnel Capacity:
1000 mL
Pore Size:
0.1 μm
Price (SGD)
1,458.00
PK
Add to cart
  • Sterile vacuum filtration system size 1L (1000 mL) with 45 mm neck size
  • Funnel contains 79 x 79 mm square membrate filter with a capacity of either 500 mL or 1000 mL
  • Receiver easily unscrews from unit and can be sealed with a plug cap for filtrate storag
  • Low liquid retention funnel design
  • Filters feature printing on the funnel for easy product identification
  • Angled hose connector simplifies vacuum line attachment
  • Receiver bottles feature easy grip sides for improved handling
  • Extra plastic storage bottles are available
  • Prefilters must be ordered separately
  • Individually packaged
  • Radiation sterilized and certified nonpyrogenic
  • Caps for receiver bottles are sterile and individually packaged
  • Units are color coded by membrane type and labeled with both membrane material and pore size
  • All membrane types are tissue culture compatible, and heat-sealed to the support grid to maximize flowrate, and reduce foaming and protein denaturization
  • Cellulose Acetate (CA) membrane for low protein binding
  • Nylon membrane for low extractables
  • Cellulose nitrate (CN) for general-purpose filtration
  • Polyethersulfone (PES) for fast flowrates and low protein binding
  • Nylon, Cellulose Nitrate and Polyetersulfone (PES) membranes are detergent-free
  • Not for parenteral use
Specifications
ConnectorsAngled Hose
DescriptionIncludes square membrane filter, graduated funnel and receiver of clear polystyrene, polyethylene neck adapter, orange HDPE plug seal
DisposableSingle-use
For Use With (Equipment)Bottle
MaterialPolystyrene
Material (Membrane)PES
Pore Size0.1 μm
SterilitySterile
Filtration Area54.5 cm2
For Use With (Application)PES membranes provide the fastest flow rates, the lowest protein binding and low extractables and are best for filtering cell culture media
GraduationsYes
Quantity12/Cs.
TypeVacuum Filter System
Volume (Metric)1000 mL
Volume (Metric) Receiver1000 mL
Funnel Capacity1000 mL
Unit SizePK