Polypropylene Wide Mouth Bottles Series 303
Polypropylene Wide Mouth Bottles Series 303
Actual product may vary
Fisher Catalogue
Polypropylene Wide Mouth Bottles Series 303
Manufactured with polypropylene for high chemical and thermal resistance. Kautex™ Polypropylene Wide Mouth Bottles are ideal for pastes, granulates and hot filled products.
Have Questions?
Catalog NumberCapacity (Metric)Neck ID (Metric)
FKS11361994100 mL24.3 mm
Catalog number FKS11361994
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
100 mL
Neck ID (Metric):
24.3 mm
  • Autoclavable up to 121° C
  • Graduated with blue ink for optimum visibility
Note: Before autoclaving screw off the closure and place it upside down on the bottle.
Application:
  • dispensing
  • sending samples
  • mailing
  • transport
  • storage
Specifications
Capacity (Metric)100 mL
ColorTranslucent
For Use With (Application)Dispensing, Sending Samples, Mailing, Transport, Storage
FormRound
IncludesClosure
MaterialPolypropylene
Neck ID (Metric)24.3 mm
Replacement ClosureScrew Closure PP
Quantity20 Pk.
AutoclavableAutoclavable
GraduatedYes
Height (Metric)94 mm
Product TypeWide Mouth Bottle
Closure TypeScrew Cap
Unit SizePK