Whatman™ Lens Cleaning Tissue
Whatman™ Lens Cleaning Tissue
Actual product may vary
Fisherbrand
Whatman™ Lens Cleaning Tissue
Provides scratch-free cleaning solutions. Whatman™ Lens Cleaning Tissue safely removes surface moisture and grease.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)Quantity
FKS11360654150 x 100 mm25 Sheets/Wallet, 25 Wallets/Cs.
FIS08-777-100150 x 100 mm25 Sheets/Wallet, 25 Wallets/Cs.
FIS09-800-973300 x 200 mm100 Sheets
FKS10149470300 x 200 mm100 Sheets
Catalog number FKS11360654
Price (SGD)
760.00
EA
Add to cart
Dimensions (L x W):
150 x 100 mm
Quantity:
25 Sheets/Wallet, 25 Wallets/Cs.
Price (SGD)
760.00
EA
Add to cart
Provides scratch-free cleaning solutions. Whatman™ Lens Cleaning Tissue safely removes surface moisture and grease.
Soft textured strong tissue leaves no fibers. Protects lenses and other optical surfaces made from glass, quartz, or plastic from scratches.
  • Chemically pure, free of silicone and other additives
  • High absorbency leads to increased safety upon removal of surface moisture and grease
  • Thickness 0.035 to 0.040mm
  • Grade 105 sheets
Specifications
Product TypeLens Wipe
DescriptionWon't damage lenses or optical surfaces, leaves no fibers
Dimensions (L x W)150 x 100 mm
For Use With (Application)Safe removal of surface moisture and grease.
Quantity25 Sheets/Wallet, 25 Wallets/Cs.
Unit SizeEA