Whatman™ 13mm Puradisc PTFE Syringe Filters
Whatman™ 13mm Puradisc PTFE Syringe Filters
Actual product may vary
Fisher Catalogue
Whatman™ 13mm Puradisc PTFE Syringe Filters
Designed to be resistant to organic solvents. Whatman™ 13mm Puradisc PTFE Syringe Filters are ideal for the clarification of aggressive organic solvents and can also be used as a venting filter for small vessels.
Have Questions?
Catalog NumberPore Size
FKS113593640.2 μm
Catalog number FKS11359364
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Pore Size:
0.2 μm
  • Hydrophobic PTFE membrane is laminated with polypropylene
  • Mini-Tip outlet is designed for filtration into very small sample bottles
  • Extremely low dead volume <10μL
  • Optimal utilization of small sample volumes
  • Excellent chemical resistance against standard organic HPLC solvents
  • Polypropylene support
  • Used as a venting filter for small vessels
Specifications
Color CodeWhite
MaterialPTFE
Pore Size0.2 μm
Housing MaterialPolypropylene
Inlet Stylefemale Luer-Lok
Outlet StyleMini tip
Quantity100/Pk.
TypeSyringe Filter
Diameter (Metric)13 mm
Unit SizePK