Sterilin™ Black and White Microtiter™ Plates
Sterilin™ Black and White Microtiter™ Plates
Actual product may vary
Fisherbrand
Sterilin™ Black and White Microtiter™ Plates
Perform both fluorescence work and bioluminescence studies with 96-well Thermo Scientific™ Sterilin™ Black and White Microtiter™ plates.
Have Questions?
Catalog NumberColor
FKS11359163Black
Catalog number FKS11359163
Price (SGD)
647.00
PK
Add to cart
Color:
Black
Price (SGD)
647.00
PK
Add to cart
Perform both fluorescence work and bioluminescence studies with 96-well Thermo Scientific™ Sterilin™ Black and White Microtiter™ plates. Supplied with no lid; if required, the 642000 lid is a suitable fit.
Features:
  • Black 96-well plates are ideal for fluorescence work where the pigment prevents cross-talk between the wells— low background fluorescence minimizes light scattering
  • White 96-well plates are ideal for bioluminescence studies where the pigment aids reflectivity and sensitivity
  • SBS compliant, ensuring suitable fit with all automation
  • Manufactured from virgin polystyrene
  • Supplied nonsterile, providing a medium binding surface
  • Chimney well design to help reduce contamination
Ordering Information:
Supplied without a lid; if required, the 642000 lid is a suitable fit.
Specifications
FormatStandard
Array8 x 12
Packaging50 Cases, 5 Pk.
ColorBlack
Product TypeMicroplate
MaterialPolystyrene
Volume (Metric) Well400 μL
No. of Wells96
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeFlat Bottom (F-well)
No. per Case50
No. per Pack5
Quantity50/Pk.
Unit SizePK