Scientific Shaker Universal Platforms
Scientific Shaker Universal Platforms
Actual product may vary
Fisherbrand
Scientific Shaker Universal Platforms
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
Have Questions?
Catalog NumberLength (English) PlatformWidth (English) PlatformFor Use With (Equipment)
FKS1135599430 in.18 in.Innova 43/43R, 2300, series 4300 and 4400, E10, E25, E25R, C10, C25/C25KC shaker
Catalog number FKS11355994
Price (SGD)
6,470.00
EA
Add to cart
Length (English) Platform:
30 in.
Width (English) Platform:
18 in.
For Use With (Equipment):
Innova 43/43R, 2300, series 4300 and 4400, E10, E25, E25R, C10, C25/C25KC shaker
Price (SGD)
6,470.00
EA
Add to cart
  • Accept a variety of accessory clamps, test tube racks and gel trays for greater flexibility
  • Hold combinations of flasks or test tube racks
  • With pre-drilled holes
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
Specifications
Product TypeUniversal Platform
MaterialAluminum
Dimensions (L x W)18 x 30 in. (457 x 762 mm)
Length (English)18 in.
Length (English) Platform30 in.
Length (Metric)457 mm
Length (Metric) Platform76 cm
Width (English)30 in.
Width (English) Platform18 in.
Width (Metric)762 mm
Width (Metric) Platform46 cm
For Use With (Equipment)Innova 43/43R, 2300, series 4300 and 4400, E10, E25, E25R, C10, C25/C25KC shaker
IncludesPre-drilled holes
Unit SizeEA