Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters
Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters
Actual product may vary
Fisher Catalogue
Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters
Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters may be used for biological sample and HPLC aqueous sample preparation.
Have Questions?
Catalog NumberPore SizeQuantity
FKS113546940.45 μm200/Pk.
Catalog number FKS11354694
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Pore Size:
0.45 μm
Quantity:
200/Pk.
Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters may be used for biological sample and HPLC aqueous sample preparation.
  • Diameter: 25mm
  • Sample volume up to 100mL
  • Low hold-up volumes for maximum sample recovery
  • Optional glass microfiber
  • Autoclave at 121°C
Applications:
  • HPLC aqueous sample preparation
  • Biological sample preparation
  • Buffer solutions
  • Salt solutions
  • Tissue culture media
  • Irrigation solutions
  • Sterile isolation
Specifications
Material (Membrane)Polypropylene Depth Filter
Pore Size0.45 μm
SterilityNon-sterile
Filtration Area4.2 cm2
Housing MaterialPigment Free Polypropylene (PP)
Inlet StyleFemale Luer Lock
Outlet StyleMale Luer
Pressure Rating5.2 bar (75 psi)
Quantity200/Pk.
TypeSyringe Filter
Volume (Metric)Up to 100 mL
Dimensions (L x W x H)22.9 x 28.4 mm
Diameter (Metric)25 mm
Unit SizePK