M6 Male Tubing Connectors
M6 Male Tubing Connectors
Actual product may vary
Fisherbrand
M6 Male Tubing Connectors
M6 tubing connectors constructed of durable plastic materials for use in chromatography pump systems. Cytiva M6 Tubing Connectors are available in male flanged, flangeless and end fitting connections.
Have Questions?
Catalog NumberConnections
FKS11350052M6 Male Flangeless
Catalog number FKS11350052
Price (SGD)
437.00
PK
Add to cart
Connections:
M6 Male Flangeless
Price (SGD)
437.00
PK
Add to cart
To prevent air from entering a chromatography system pump, tubing connections at the pump and inlets must be tight and secure.
  • Male Connector, Flangeless 
  • Male Connector: I.D. 3.3mm
  • Male End fitting: I.D. 2mm
Specifications
TypeTubing Connector
ConnectionsM6 Male Flangeless
Quantity2 Pk.
MaterialFPM, Polypropylene, POM
For Use With (Equipment)Chromatography Pump Systems, Tubing
Unit SizePK