Parafilm™ Sealing Film Dispenser
Parafilm™ Sealing Film Dispenser
Actual product may vary
Fisherbrand
Parafilm™ Sealing Film Dispenser
Cuts and dispenses film safely. Parafilm™ Sealing Film Dispenser is supplied with two safety razor cutting blades.
Have Questions?
Catalog NumberColor
FKS11350040Blue
Catalog number FKS11350040
Price (SGD)
209.00
EA
Add to cart
Color:
Blue
Price (SGD)
209.00
EA
Add to cart
  • ABS plastic dispenser designed with a safety razor that slides and cuts with no danger to fingers.
  • Supplied with two easily exchanged cutting blades.
  • Dispenser holds one or two 50 mm (2-inch) rolls, or one 100 mm (4-inch) roll.
Specifications
Product TypeSealing Film Dispenser
ColorBlue
Length (English)6.7 in.
Length (Metric)171 mm
Width (English)4.7 in.
Width (Metric)120 mm
Height (English)6.1 in.
Height (Metric)156 mm
HoldsParafilm
MaterialABS
MountingFreestanding
No. of Compartments1
Product StyleHinged Lid
Capacity1 x 100 mm roll or 2 x 50 mm rolls
Includes2 cutting blades
Quantity1 Ea.
Unit SizeEA