Azlon™ Multi-lingual Safety Venting Wash Bottle
Azlon™ Multi-lingual Safety Venting Wash Bottle
Actual product may vary
Fisher Catalogue
Azlon™ Multi-lingual Safety Venting Wash Bottle
Store and use a variety of solvents with multi-lingual labels for improved safety. Azlon Multi-lingual LDPE Venting Wash Bottles are available in four different multi-language labeling combinations. Solvent content labels correspond to color-coded lids for easy identification in a variety of settings.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Chemical LabelDescriptionClosure Color
FKS11349643500 mLEthanolPrinted in English, French, German and SpanishOrange
FKS11329643500 mLAcetonePrinted in English, French, German and SpanishRed
FKS11303524500 mLIsopropanolPrinted in English, French, German and SpanishBlue
Catalog number FKS11349643
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
500 mL
Chemical Label:
Ethanol
Description:
Printed in English, French, German and Spanish
Closure Color:
Orange
Store and use a variety of solvents with multi-lingual labels for improved safety. Azlon Multi-lingual LDPE Venting Wash Bottles are available in four different multi-language labeling combinations. Solvent content labels correspond to color-coded lids for easy identification in a variety of settings.
  • Multi-lingual wide neck wash bottles featuring the patented DripLok™ vapor-venting valve that prevents solvent drips
  • Indelibly printed in multiple languages (based on region)
  • Includes the Globally Harmonised Symbols, NFPA diamond, risk phrases, and safety information
  • Fitted with color coded one piece PP closures that maintain their shape in use
  • Closures feature precision jet-tip aperture for fine control
Specifications
Capacity (Metric)500 mL
Capacity (English)16.91 oz.
ColorTranslucent
DescriptionPrinted in English, French, German and Spanish
IncludesPolypropylene caps and colored beaks with fine nozzle end to control the jet accurately
MaterialLDPE
Length (Metric)33.07cm
Quantity5/PK
Temperature Range (Metric)–50 to 80°C
Width (Metric)19.08cm
Closure ColorOrange
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthWide
ShapeCylindrical
Height (Metric)5.6 cm
Product TypeMulti-Lingual Printed Wash Bottle
Chemical LabelEthanol
Closure MaterialPolypropylene
Closure TypeCap
Through the Closure DispensingYes
Unit SizePK