Quickfit™ Standard Taper Flat Head Borosilicate Glass Hollow Stopper
Quickfit™ Standard Taper Flat Head Borosilicate Glass Hollow Stopper
Actual product may vary
Quickfit™ Standard Taper Flat Head Borosilicate Glass Hollow Stopper
Use this glass stopper with Quickfit™ jointed glassware and other glassware having standard ground glass sockets. Azlon™ Standard Taper Flat Head Borosilicate Glass Hollow Stoppers are lightweight, durable and easy to grip.
Have Questions?
Catalog NumberStandard Taper Joint
FKS11349623Standard; 34/35
Catalog number FKS11349623
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Standard Taper Joint:
Standard; 34/35
Use this glass stopper with Quickfit™ jointed glassware and other glassware having standard ground glass sockets. AzlonTM Standard Taper Flat Head Borosilicate Glass Hollow Stoppers are lightweight, durable and easy to grip.
  • Conforms to the standard tapers as outlined in ISO 383
  • Borosilicate glass for chemical durability and high temperature performance
  • Hollow-blown construction to reduce weight
  • Hexagonal head prevents the stopper from rolling and provides a good grip
  • Top of stopper is flat, allowing them to be stood on end and avoid contamination of the ground surface
Specifications
For Use With (Application)Chemical durability and high temperature performance
For Use With (Equipment)Quickfit™ jointed ware and other Glassware having standard ground Glass sockets
Standard Taper JointStandard; 34/35
MaterialBorosilicate Glass
Product TypeStopper
Unit SizePK